Merck
  • [A method for the recovery of dioxane, naphthalene, PPO and POPOP from used scintillation solutions].
[A method for the recovery of dioxane, naphthalene, PPO and POPOP from used scintillation solutions].
Zeitschrift fur medizinische Laboratoriumsdiagnostik (1987-01-01)
G Merkel, P Lange
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, BioReagent, suitable for scintillation