Merck
Kinetics of sorption of ionizable solutes by plastic infusion bags.
Journal of pharmaceutical sciences (1986-06-01)
E A Kowaluk, M S Roberts, A E Polack
ABSTRACT
Aqueous solutions of several ionizable substances were stored in plastic infusion bags and the sorption of the substances monitored with time. The substances used were p-nitrophenol, p-toluidine, warfarin sodium [3-(alpha-acetonylbenzyl)-4-hydroxycoumarin sodium salt] and trifluoperazine hydrochloride (10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)phenothiazine dihydrochloride). The rate and extent of sorption for each substance varied with pH and was consistent with a preferential uptake of the un-ionized species. The uptake of p-nitrophenol and p-toluidine was adequately described by a diffusion model derived assuming that sorption is rate-controlled by the diffusivity of the solute in the plastic matrix, and that only the un-ionized species was sorbed by the plastic matrix. However, the uptake of warfarin sodium and trifluoperazine hydrochloride was described more accurately by a diffusion model in which the diffusional resistance of the plastic matrix and of an interfacial resistance barrier both contributed to the diffusional resistance encountered in the sorption process. It appeared that the rate of uptake of the un-ionized form of these solutes was diminished due to the influence of interfacial or aqueous diffusional barriers. Solute lipophilicity and degree of ionization appeared to be important factors determining the relative contribution of the respective barriers to the overall diffusional resistance.
Sigma-Aldrich
p-Toluidine, 99%
Sigma-Aldrich
p-Toluidine hydrochloride, purum, ≥99.0% (AT)
Sigma-Aldrich
p-Toluidine, 99.6%
Supelco
p-Toluidine, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%