Merck
[Cysteine stability in aqueous amino acid solutions].
Prikladnaia biokhimiia i mikrobiologiia (1984-05-01)
A D Nekliudov, I V Verem'ëv
ABSTRACT
The effect of pH on the cysteine content in the model solution containing 17 L-amino acids and D-sorbitol, as well as the effect of traditional antioxidants potassium metabisulfite and ascorbic acid on the cysteine stability were studied. Optimal conditions for cysteine stabilization in aqueous solutions of amino acids, available for parenteral protein feeding, were found.
Sigma-Aldrich
Potassium disulfite, BioUltra, ≥97.0% (RT)
Sigma-Aldrich
Potassium disulfite, ≥98%
Sigma-Aldrich
Potassium disulfite, puriss., meets analytical specification of Ph.Eur., BP, NF, FCC, E224, 95.0-101.0% (iodometric), powder, 51.8-57.6% sulfur dioxide