Merck
  • Carcinogenicity tests of certain environmental and industrial chemicals.
Carcinogenicity tests of certain environmental and industrial chemicals.
Journal of the National Cancer Institute (1981-07-01)
E K Weisburger, B M Ulland, J Nam, J J Gart, J H Weisburger
ABSTRACT
Fourteen chemicals of varied uses were tested for carcinogenicity by oral administration in male and female Charles River CD rats. Under the conditions of the tests, propane sultone, propylene imine, and ethylenethiourea, in addition to the positive control N-2-fluorenylacetamide, were carcinogenic. Avadex, bis(2-chloroethyl) ether, the potassium salt of bis(2-hydroxyethyl) dithiocarbamic acid, ethylene carbonate, and semicarbazide hydrochloride were not carcinogenic under the test conditions. Dithiooxamide, glycerol alpha-monochlorohydrin, and thiosemicarbazide gave somewhat ambiguous results, though administered at high enough dose levels to be toxic. An inadequate number of animals survived treatments with sodium azide, sodium bisulfide, and vinylene carbonate, or the animals may not have received sufficiently high doses of the test chemicals to provide maximum test sensitivity. However, there were no indications that these three chemicals were carcinogenic under the test conditions.
Sigma-Aldrich
2-Chloroethyl ether, 99%
Sigma-Aldrich
Dithiooxamide, 97%
Sigma-Aldrich
Bis(2-chloroethyl) ether, puriss., ≥99.0% (GC)
Sigma-Aldrich
1,3-Propanesultone, 98%
Sigma-Aldrich
1,3-Propanesultone, ≥99%
Sigma-Aldrich
Ethylene carbonate, 98%
Sigma-Aldrich
Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%