Merck
  • Chlorodecarboxylation of 17 beta-acetoxy-3-methoxy-9-oxo-9, 11-secoestra-1, 3, 5 (10)-trien-11-oic acid with lead tetraacetate and trityl chloride.
Chlorodecarboxylation of 17 beta-acetoxy-3-methoxy-9-oxo-9, 11-secoestra-1, 3, 5 (10)-trien-11-oic acid with lead tetraacetate and trityl chloride.
Steroids (1982-05-01)
K Lal, S Ray
Sigma-Aldrich
Trityl chloride, 97%
Sigma-Aldrich
Trityl chloride, purum, ≥97.0% (HPLC), ≥97.0% (AT)