Merck
Chlordane sampling efficiency with Chromosorb 102 and ethylene glycol.
American Industrial Hygiene Association journal (1980-08-01)
T C Thomas, J W Jackson, Y A Nishioka
ABSTRACT
An organic polymeric solid, Chromosorb 102, has been reported to collect various pesticides from air. In this study the collection efficiency of Chromosorb 102 for chlordane was evaluated. Preparation of sampling tubes containing this medium for field sampling is also discussed.
Product Description
Supelco
Poly(styrene-co-divinylbenzene), microspheres, 6.0-10.0 μm avg. part. size
Pricing and availability is not currently available.