Merck
  • Preparation, characterization and pharmaceutical application of linear dextrins. I. Preparation and characterization of amylodextrin, metastable amylodextrins, and metastable amylose.
Preparation, characterization and pharmaceutical application of linear dextrins. I. Preparation and characterization of amylodextrin, metastable amylodextrins, and metastable amylose.
Pharmaceutical research (1993-09-01)
G H Te Wierik, A C Eissens, A C Besemer, C F Lerk
ABSTRACT
The linear dextrin amylodextrin was prepared by enzymatic hydrolysis from waxy maize. Four metastable amylodextrins were prepared by complexation with different volatile organic compounds. All products showed partial dissolution into water at room temperature, because of dissolution of molecules with a lower DP. X-ray diffractometry revealed a helical conformation with six glucose units per turn for amylodextrin and metastable amylodextrins prepared with small molecules, and a helical conformation with seven glucose units per turn for metastable amylodextrins prepared with larger molecules. All metastable amylodextrins showed a helix with reduced distance between two turns as compared to amylodextrin. Metastable amylose, prepared from Amylose V, showed a helical conformation again with a reduced distance between two turns compared to Amylose V. FTIR analysis indicated a more flexible conformation for Amylose V and metastable amylose than for the amylodextrins.
Sigma-Aldrich
Starch, soluble, ACS reagent