Merck
  • Protriptyline as an alternative stimulant medication in patients with brain injury: a series of case reports.
Protriptyline as an alternative stimulant medication in patients with brain injury: a series of case reports.
Brain injury (1993-07-01)
B Wroblewski, M B Glenn, R Cornblatt, A B Joseph, S Suduikis
ABSTRACT
The results of a series of eight individual case reports in which protriptyline, an activating tricyclic antidepressant, was used as a 'stimulant' medication are presented. For some patients with head injury, traditional stimulants, such as methylphenidate, or dopaminergic stimulants, such as levodopa-carbidopa, amantadine, or bromocriptine, may be partially or totally ineffective or not tolerated. Protriptyline can be a very effective alternative and, for some patients, may be the most effective stimulant tried. In low to moderate doses, protriptyline should be considered for trials as an activating/stimulant medication in patients with head injury.
Product Description
Supelco
Protriptyline hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®