Merck
Differentiation of esterases reacting with organophosphorus compounds.
Chemico-biological interactions (1993-06-01)
E Reiner, E Pavković, Z Radić, V Simeon
ABSTRACT
The hydrolysis of paraoxon (POX), phenylacetate (PA) and beta-naphthylacetate (BNA) was studied in human serum. Based upon correlations between enzyme activities, upon reversible inhibition by EDTA and upon progressive inhibition by iso-OMPA, tabun, eserine and bis-4 nitrophenylphosphate, the following conclusions were drawn about the number and specificity of enzymes involved in the hydrolysis. Two paraxonases hydrolyse paraoxon: one sensitive and the other insensitive to EDTA. The EDTA-sensitive paraoxonase also hydrolysed BNA. The EDTA-insensitive hydrolysis of BNA and PA was attributed to a serine esterase. The EDTA-sensitive hydrolysis of PA is probably due to more than one enzyme, which might be an arylesterase and a carboxylesterase.
Sigma-Aldrich
Propionylthiocholine iodide, ≥98%
Sigma-Aldrich
2-Naphthaleneacetic acid, 99%