Merck
  • Determination of UV stabilizers in PET bottles by high performance-size exclusion chromatography.
Determination of UV stabilizers in PET bottles by high performance-size exclusion chromatography.
Food additives and contaminants (1996-07-01)
M Monteiro, C Nerín, F G Reyes
ABSTRACT
A Size Exclusion Chromatography-High Performance Liquid Chromatography (SECHPLC) method to determine antioxidants and UV stabilizers in PET bottles has been developed. In only a single run a synthetic mixture of the stabilizers was separated and quantified. The detection limit obtained for BHT, Tinuvin 326, Cyasorb UV 5411, and Tinuvin P was about 0.1 microgram/g and for Irgafos 168 it was 1.0 microgram. RSD values were lower than 3%. Tinuvin P was identified and quantified in PET bottle extracts. Olive oil, soybean oil and sunflower oil showed well defined separation from Tinuvin P at the same conditions of analysis. Cyclic dimers were identified in the PET extracts.
Product Description
Plastic additive 12, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-(2-Hydroxy-5-methylphenyl)benzotriazole, 97%
Sigma-Aldrich
2-tert-Butyl-6-(5-chloro-2H-benzotriazol-2-yl)-4-methylphenol, 98%
Sigma-Aldrich
Tris(2,4-di-tert-butylphenyl) phosphite