Merck
  • Is local biotransformation the key to understanding the pharmacological activity of salicylates and gold drugs?
Is local biotransformation the key to understanding the pharmacological activity of salicylates and gold drugs?
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (1996-12-01)
M W Whitehouse, G G Graham
ABSTRACT
It is suggested that some drugs may be converted by inflammatory cells to yield active species. The transformation may be non-enzymatic, although being driven by the enzymatic production of highly reactive species which are normal products of activated leukocytes, such as singlet oxygen, hydrogen peroxide, hypochlorite, hydroxyl radical and nitric oxide. Drugs which may be transformed in this fashion are the anti-rheumatic gold complexes which may be converted either to aurocyanide or to Au(III) complexes by myeloperoxidase in polymorphonuclear leukocytes. Salicylate may also be activated by its oxidation to dihydroxybenzoates although evidence for its transformation is weaker than for the gold complexes.
Sigma-Aldrich
Potassium dicyanoaurate(I), 99.95% trace metals basis
Sigma-Aldrich
Potassium dicyanoaurate(I), 98%