Merck
  • Metanil yellow: a bifunctional inducer of hepatic phase I and phase II xenobiotic-metabolizing enzymes.
Metanil yellow: a bifunctional inducer of hepatic phase I and phase II xenobiotic-metabolizing enzymes.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1997-08-01)
M Das, S Ramchandani, R K Upreti, S K Khanna
ABSTRACT
Metanil yellow, a non-permitted food colour, has been found in various foodstuffs. The induction potential of metanil yellow on hepatic microsomal cytochrome P-450 (P-450)-dependent monooxygenases and cytosolic detoxification enzymes, namely, glutathione S-transferase (GST) and quinone reductase (QR), was investigated. Oral administration of metanil yellow (430 mg/kg body weight) to four animals for seven days caused significant induction of hepatic P-450 (48%) and its dependent aryl hydrocarbon hydroxylase (100%) activity and cytosolic GST (136%) and QR (92%) activities. Parenteral administration of metanil yellow (80 mg/kg body weight) to another set of four animals for 3 days resulted in higher induction of ethoxyresorufin-O-deethylase (228%) as compared to other monooxygenases (64-92%), while GST and QR were also found to be induced (59-95%). Spectra of metanil yellow-induced microsomes showed an increase in P-450 with a shift of 2.2 nm in the soret region. The results suggest that metanil yellow acts as a bifunctional inducer of specific isozymes of P-450 and cytosolic enzymes and thus may involve the cytosolic aryl hydrocarbon (Ah) receptor for this type of induction.
Sigma-Aldrich
Metanil Yellow, Dye content 70 %
Supelco
Metanil Yellow, analytical standard