Merck
  • Midazolam can potentiate the analgesic effects of intrathecal bupivacaine on thermal- or inflammatory-induced pain.
Midazolam can potentiate the analgesic effects of intrathecal bupivacaine on thermal- or inflammatory-induced pain.
Anesthesia and analgesia (2003-04-23)
Tomoki Nishiyama, Kazuo Hanaoka
ABSTRACT
Epidurally administered midazolam can potentiate analgesia by epidural bupivacaine. However, whether this effect is synergistic or additive is not known. In this study, we investigated the spinally-mediated analgesic interaction between midazolam and bupivacaine by using the tail-flick and formalin tests in rats with chronically implanted catheters. Behavioral effects were also observed. The dose dependency of analgesia and the 50% effective doses of intrathecal midazolam and bupivacaine were determined, and then the interaction of these two drugs was examined with an isobolographic analysis. Both drugs had dose-dependent analgesic effects in both the tail-flick test and the formalin test. The 50% effective dose values of the combination were significantly lower than the calculated additive values in both tests (P = 0.023 in the tail-flick test; P = 0.0025 in Phase 1 and 0.047 in Phase 2 of the formalin test). Behavioral side effects decreased in the combination group compared with each drug alone. In conclusion, intrathecally administered midazolam and bupivacaine had synergistic analgesic effects on acute thermal- or inflammatory-induced pain, with decreased behavioral side effects. In both acute thermal- and inflammatory-induced pain, intrathecally administered midazolam and bupivacaine produced synergistic analgesia with decreased side effects in intrathecally catheterized rats.