Merck
  • A review of analytical methods for the determination of sulfolane and alkanolamines in environmental studies.
A review of analytical methods for the determination of sulfolane and alkanolamines in environmental studies.
Journal of AOAC International (2002-03-07)
John V Headley, Phillip M Fedorak, Leslie C Dickson
ABSTRACT
Sulfolane and alkanolamines are used extensively in the processing of sour natural gases. Over many years of operation, there have been inadvertent leaks of these chemicals to groundwater and wetlands surrounding gas processing facilities, leading to uptake by vegetation. Because sulfolane and alkanolamines are extremely water-soluble, their analysis has presented challenges, particularly requirements for suitable extraction from biological matrixes and soil, along with sensitive detection using commonly available instrumentation. Analytical methods usually use gas chromatography or liquid chromatography with a variety of detector systems. Sample preparation techniques may include extraction with organic solvents, water, or a combination of these. In some cases, direct aqueous injections have been used. Derivatization of alkanolamines has been used to improve the chromatographic separations and detection. More recent procedures, using positive-ion electrospray ionization mass spectrometry (MS), have been useful for the confirmation of uptake of the alkanolamines and transformation products by wetland vegetation. Future developments will likely center on further MS analyses for identification of metabolites and transformation products in aquatic environments.
Sigma-Aldrich
Sulfolane, 99%
Supelco
Sulfolane, analytical standard