Merck
  • [Lerkanidipin: evolution of dihydropyridine calcium antagonists continues].
[Lerkanidipin: evolution of dihydropyridine calcium antagonists continues].
Kardiologiia (2012-12-15)
V V Fomin, A A Svistunov
ABSTRACT
We discuss the clinical studies on the efficacy and tolerability of lerkanidipine in arterial hypertension.
Sigma-Aldrich
Lercanidipine hydrochloride, ≥98% (HPLC)