Merck
A fluorometric assay for biotinidase.
Analytical biochemistry (1986-04-01)
H Ebrahim, K Dakshinamurti
ABSTRACT
An assay for biotinidase using biocytin, the natural substrate, is described. The fluorometric procedure uses 1,2-diacetylbenzene which reacts selectively with lysine allowing its direct determination in mixture with biocytin. We have examined the applicability of the assay using human serum biotinidase.
Sigma-Aldrich
1,2-Diacetylbenzene, 99%