Merck
  • Forced-flow bioreactor for sucrose inversion using ceramic membrane activated by silanization.
Forced-flow bioreactor for sucrose inversion using ceramic membrane activated by silanization.
Biotechnology and bioengineering (1989-02-20)
M Nakajima, A Watanabe, N Jimbo, K Nishizawa, S Nakao
ABSTRACT
A forced-flow enzyme membrane reactor system for sucrose inversion was investigated using three ceramic membranes having different pore sizes. Invertase was immobilized chemically to the inner surface of a ceramic membrane activated by a silane-glutaraldehyde technique. With the cross-flow filtration of sucrose solution, the reaction rate was a function of the permeate flux, easily controlled by pressure. Using 0.5 microm support pore size of membrane, the volumetric productivity obtained was 10 times higher than that in a reported immobilized enzyme column reactor, with a short residence time of 5 s and 100% conversion of the sucrose inversion.