Merck
  • METI-acaricide resistance in Tetranychus urticae does not confer resistance to the naphthoquinones.
METI-acaricide resistance in Tetranychus urticae does not confer resistance to the naphthoquinones.
Pest management science (2001-08-24)
G J Devine, B P Khambay
ABSTRACT
The naphthoquinones and the METI group of compounds act on sites associated with mitochondrial respiration, but METI-resistant strains of two-spotted spider mite from Japan and the UK exhibited no cross-resistance to the naphthoquinones. The potential for developing commercial naphthoquinones therefore remains high.