Merck
  • Inhibition of prostalglandin E2 production by 2'-hydoxychalcone derivatives and the mechanism of action.
Inhibition of prostalglandin E2 production by 2'-hydoxychalcone derivatives and the mechanism of action.
The Journal of pharmacy and pharmacology (2001-10-02)
Y P Kim, H S Ban, S S Lim, N Kimura, S H Jung, J Ji, S Lee, N Ryu, S R Keum, K H Shin, K Ohuchi
ABSTRACT
The effects of 14 synthetic 2'-hydroxychalcone derivatives on prostaglandin E2 (PGE2) production in rat peritoneal macrophages stimulated by the protein kinase C activator, 12-O-tetradecanoylphorbol 13-acetate (TPA), were examined to clarify the structure-activity relationship. 2',4-Dihydroxy-4'-methoxychalcone (compound 3), 2',4-dihydroxy-6'-methoxychalcone (compound 8) and 2'-hydroxy-4'-methoxychalcone (compound 9) suppressed PGE2 production more potently than the other compounds. The IC50 (50% Inhibitory concentration) value for compounds 3, 8 and 9 was calculated to be 3 microM. The activity of cyclooxygenase (COX)-1 was inhibited slightly by compound 9, but that of COX-2 was not inhibited. At concentrations that inhibited the production of PGE2, compound 9 had no effect on the release of radioactivity from [3H]arachidonic acid-labelled macrophages stimulated by TPA. Western-blot analysis revealed that the induction of COX-2 protein by TPA was inhibited by compound 9 in parallel with the inhibition of PGE2 production. Compounds 3 and 8 had similar effects. These findings suggest that 4'-methoxyl and 6'-methoxyl groups are required for the expression of more potent inhibitory activity against PGE2 production, and that the inhibition of PGE2 production by these 2'-hydroxychalcone derivatives is due to the inhibition of TPA-induced COX-2 protein expression.