Merck
  • Persistence and residual activity of an organophosphate, pirimiphos-methyl, and three IGRs, hexaflumuron, teflubenzuron and pyriproxyfen, against the cowpea weevil, Callosobruchus maculatus (Coleoptera: Bruchidae).
Persistence and residual activity of an organophosphate, pirimiphos-methyl, and three IGRs, hexaflumuron, teflubenzuron and pyriproxyfen, against the cowpea weevil, Callosobruchus maculatus (Coleoptera: Bruchidae).
Pest management science (2004-01-20)
Gamal E Abo-Elghar, Anwar E El-Sheikh, Ferial M El-Sayed, Hamdi M El-Maghraby, Hesham M El-Zun
ABSTRACT
Three insect growth regulators (IGR), the chitin synthesis inhibitors (CSI) teflubenzuron and hexaflumuron and the juvenile hormone mimic (JHM) pyriproxyfen, as well as the organophosphate (OP) pirimiphos-methyl, were evaluated for their activity against the cowpea weevil, Callosobruchus maculatus (F), in cowpea seeds stored for up to 8 months post-treatment. The initial activity data showed that, based on LC50 level, teflubenzuron had strong ovicidal activity (LC50 = 0.056 mg kg(-1)) followed by pirimiphos-methyl (1.82 mg kg(-1)) and pyriproxyfen (91.9 mg kg(-1)). The residual activity data showed that none of the IGRs tested had strong activity when applied at 200 mg kg(-1) in reducing the oviposition rates of C maculatus at various storage intervals up to 8 months post-treatment. However, teflubenzuron reduced adult emergence (F1 progeny), achieving control ranging from 96.2% at 1 month to 94.3% at 8 months. Hexaflumuron showed a similar trend in its residual activity, ranging between 93.8% control at 1 month to 88.2% control at 8 months post-treatment. However, pyriproxyfen was more active than the CSIs tested and caused complete suppression (100% control) of adult emergence at all storage intervals. Unlike the IGRs tested, pirimiphos-methyl applied at 25 mg kg(-1) was more effective in reducing oviposition rates of C maculatus up to 8 months post-treatment. A strong reduction of adult emergence was also observed at various bimonthly intervals (98.6% control at 1 month to 91.6% control at 8 months post-treatment). The persistence of hexaflumuron and pirimiphos-methyl in cowpea seeds was also studied over a period of 8 months. The loss of hexaflumuron residue in treated cowpeas (200 mg kg(-1)) was very slow during the first month post-treatment (4.43%). At the end of 8 months, the residue level had declined significantly to 46.4% of the initial applied rate. The loss of pirimiphos-methyl residue in treated cowpeas (25 mg kg(-1)) was relatively high during the first month post-treatment (36.7%) and increased to 81.6% after 8 months.
Product Description
Supelco
Teflubenzuron, PESTANAL®, analytical standard
Supelco
Hexaflumuron, PESTANAL®, analytical standard