Merck
Extended follow-up of cancer incidence in fluoride-exposed workers.
Journal of the National Cancer Institute (2004-05-20)
Philippe Grandjean, Jørgen H Olsen
Sigma-Aldrich
Cryolite, synthetic, ≥97.0% (from F)