Merck
  • Evaluation of pheromone release from commercial mating disruption dispensers.
Evaluation of pheromone release from commercial mating disruption dispensers.
Journal of agricultural and food chemistry (2005-03-31)
Elizabeth Tomaszewska, Vincent R Hebert, Jay F Brunner, Vincent P Jones, Mike Doerr, Richard Hilton
ABSTRACT
Pome fruit growers and crop consultants have expressed concerns about the seasonal release performance of commercial codling moth mating disruption dispenser products. Because of these concerns, we developed a laboratory flow-through volatile collection system (VCS) for measuring the volatile release of the codling moth sex pheromone, codlemone, from commercially available hand-applied dispensers. Under controlled air-flow and temperature conditions, the released vapor was trapped onto a polyurethane foam adsorbent followed by solvent extraction, solvent reduction, and GC/MS determination. Method recovery and breakthrough validations were performed to demonstrate system reliability before determining codlemone release from commercial dispensers field-aged over 140 days. The volatile collection was carried out in a consistent manner among five dispenser types most commonly used by growers, so that direct comparison of performance could be made. The comparison showed differences in the amount of pheromone released and in the patterns of release throughout the season between dispenser types. The variation in release performance demonstrates the need for routine evaluation of commercially marketed mating disruption dispensers. We believe that the simple and cost-effective volatile collection system can assist pheromone dispenser manufacturers in determining seasonal dispenser performance before new products are introduced into the commercial market and in rapidly verifying dispenser release when field-aged dispenser efficacy is in question.