Merck
  • Synthesis of a laterally branched polyamine from alpha-methylene-gamma-butyrolactone.
Synthesis of a laterally branched polyamine from alpha-methylene-gamma-butyrolactone.
Organic letters (2005-06-17)
Mickaël Jean, Myriam Le Roch, Jacques Renault, Philippe Uriac
ABSTRACT
[reaction: see text] A polyamine derivative was prepared from alpha-methylene-gamma-butyrolactone. This method used Michael addition and lactone aminolysis followed by the nucleophilic substitution of the hydroxyl group by an azido group. The coupling of a lipophilic alkyne led to a polyamine that will be probed as a gene transfer agent.
Sigma-Aldrich
α-Methylene-γ-butyrolactone, 97%