Merck
  • Effect of high-dose aspirin on CYP2E1 activity in healthy subjects measured using chlorzoxazone as a probe.
Effect of high-dose aspirin on CYP2E1 activity in healthy subjects measured using chlorzoxazone as a probe.
Journal of clinical pharmacology (2006-01-07)
Ji-Young Park, Kyoung-Ah Kim, Pil-Whan Park, Jong-Myung Ha
ABSTRACT
The authors evaluated the effect of high-dose aspirin at a therapeutic dose, using chlorzoxazone as a probe for CYP2E1 enzyme activity. In a randomized, open-label, 2-way crossover study, 10 healthy men were treated 3 times daily for 6 days with 1 g aspirin or placebo. On day 7, 1 dose of 400 mg chlorzoxazone was administered orally. Plasma concentrations of chlorzoxazone and its metabolite, 6-hydroxychlorzoxazone, were measured. During the aspirin phase, the area under the time-concentration curve (AUC) and peak plasma concentration of chlorzoxazone were 95% (90% confidence interval [CI], 87%-103%) and 90% (90% CI, 80%-101%) of the values during the placebo phase, respectively. High-dose aspirin did not affect the oral clearance of chlorzoxazone significantly (90% CI, 98%-120%; P = .24). The AUC ratio and plasma concentration ratios of 6-hydroxychlorzoxazone/chlorzoxazone were not changed significantly by high-dose aspirin. High-dose aspirin at a therapeutic dose does not affect CYP2E1 activity in humans.
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)