Merck
  • N-acetylmuramoyl-L-alanine amidase assay based on specific radioactive labeling of muropeptide L-alanine: quantitation of the enzyme activity in the autolysin deficient Bacillus subtilis 168, flaD strain.
N-acetylmuramoyl-L-alanine amidase assay based on specific radioactive labeling of muropeptide L-alanine: quantitation of the enzyme activity in the autolysin deficient Bacillus subtilis 168, flaD strain.
Analytical biochemistry (1991-10-01)
P Margot, C A Roten, D Karamata
ABSTRACT
A sensitive and highly reproducible assay for N-acetylmuramoyl-L-alanine amidase (EC 3.5.1.28) was devised, based on specific and homogeneous L-[14C]alanine labeling of the substrate, the peptidoglycan. The method involves partial purification of both the enzyme and the substrate and monitoring the muropeptide cleavage by coupling fluorodinitrobenzene to freed L-alanine NH2 groups. After acid hydrolysis of the substrate, the resulting DNP-L-alanine and L-alanine are separated by TLC, and radioactive counts in relevant spots are determined. Application of the method to the autolysin-endowed strain and an autolysis-deficient flaD-bearing mutant has revealed (i) that the N-acetylmuramoyl-L-alanine amidase behaves like an endoenzyme with an apparent Kcat(s-1) of 40, and (ii) that the residual enzyme activity in the flaD bearing strain amounts to 2.5 (+/- 0.1)% of that of the parent strain.