Merck
  • Rapid detection of Pneumocystis carinii in bronchoalveolar lavage samples by using Cellufluor staining.
Rapid detection of Pneumocystis carinii in bronchoalveolar lavage samples by using Cellufluor staining.
Journal of clinical microbiology (1990-02-01)
V S Baselski, M K Robison, L W Pifer, D R Woods
ABSTRACT
Cellufluor (Calcofluor white) has been found to be a useful, rapid chemofluorescent stain for detection of Pneumocystis carinii cysts in bronchoalveolar lavage samples. When compared with toluidine blue O and Giemsa stains on 45 specimens (22 positive and 23 negative), the sensitivity and specificity of the Cellufluor stain were 95 and 100%, respectively.