Merck
  • Selective binding of imatinib to the genetic variants of human alpha1-acid glycoprotein.
Selective binding of imatinib to the genetic variants of human alpha1-acid glycoprotein.
Biochimica et biophysica acta (2006-09-30)
Ilona Fitos, Júlia Visy, Ferenc Zsila, György Mády, Miklós Simonyi
ABSTRACT
Imatinib is a selective tyrosine kinase inhibitor, successfully used for the treatment of chronic myelogenous leukaemia. Its strong plasma protein binding referred to alpha1-acid glycoprotein (AGP) component was found to inhibit the pharmacological activity. AGP shows genetic polymorphism and the two main genetic variants have different drug binding properties. The binding characteristics of imatinib to AGP genetic variants and the possibility of its binding interactions were investigated by various methods. The results proved that binding of imatinib to the two main genetic variants is very different, the high affinity binding belongs dominantly to the F1-S variant. This interaction is accompanied with specific spectral changes (induced circular dichroism, UV change, intrinsic fluorescence quenching), suggesting that the bound ligand has chiral conformation that would largely overlap with other ligands inside the protein cavity. Binding parameters of Ka=1.7(+/-0.2)x10(6)M(-1) and n=0.94 could be determined for the binding on the F1-S variant at 37 degrees . Imatinib binding on the A variant is weaker and less specific. The binding affinity of imatinib to human serum albumin (nKa approximately 3 x 10(4)M(-1)) is low. Pharmacologically relevant binding interactions with other drugs can be expected on the F1-S variant of AGP.