Merck
  • Functional consequences of GABAA receptor alpha 4 subunit deletion on synaptic and extrasynaptic currents in mouse dentate granule cells.
Functional consequences of GABAA receptor alpha 4 subunit deletion on synaptic and extrasynaptic currents in mouse dentate granule cells.
Alcoholism, clinical and experimental research (2007-12-12)
Jing Liang, Asha Suryanarayanan, Dev Chandra, Gregg E Homanics, Richard W Olsen, Igor Spigelman
ABSTRACT
The alpha 4 subunit-containing gamma-aminobutyric acid (A) receptors (GABA(A)Rs) are highly expressed primarily at extrasynaptic sites in the dentate gyrus (DG) and thalamus and are suspected to contribute to tonic inhibition that is sensitive to potentiation by gaboxadol and ethanol (EtOH). Global alpha 4 subunit knockout (KO) mice exhibit greatly reduced tonic currents and insensitivity to ataxic, sedative and analgesic effects of gaboxadol compared to wild type (WT) controls. The alpha 4 KO mice were also significantly more sensitive to pentylenetetrazol-induced seizures. However, no differences were observed between alpha 4 KO and WT mice in other baseline behaviors or in the effects of EtOH on these behaviors. To examine possible functional and pharmacological GABA(A)R alterations, and search for causes for the lack of differences in EtOH behaviors we studied the effects of acute EtOH application on GABA(A)R-currents of DG cells from alpha 4 KO and WT control mice complemented by Western blot measurements. We studied the consequences of alpha 4 subunit deletion using Western immunoblotting and whole cell patch recordings from DG cells in brain slices from alpha 4 KO and WT mice. The magnitude of tonic current and its potentiation by EtOH (10 to 100 mM), alphaxalone (3 microM), and Ro15-4513 (0.3 microM) was greatly attenuated in alpha 4 KO mice. The kinetics of miniature inhibitory postsynaptic currents (mIPSCs) in alpha 4 KO mice were significantly slower compared to WT mice. Potentiation of mIPSCs by alphaxalone was greatly reduced in alpha 4 KO mice. Ro15-4513 had no effect on mIPSCs from WT or KO mice. However, mIPSCs of alpha 4 KO mice were significantly more sensitive to EtOH than those from WT mice. The gamma 2 subunit protein levels were selectively increased in hippocampus and thalamus, but not cortex of alpha 4 KO mice. These data suggest that the global loss of alpha 4 subunits leads to region- and cell location-specific compensatory increases in gamma 2 subunits, which in turn alter the pharmacological sensitivity of synaptic and extrasynaptic GABA(A)R-currents. Our data also suggests that while enhancement of tonic inhibitory currents by gaboxadol, alphaxalone, and EtOH are reduced, and behavioral sensitivity to gaboxadol and alphaxalone may be reduced, compensatory changes in synaptic GABA(A)R subunits may prevent similar reductions in behavioral sensitivity to EtOH.
Sigma-Aldrich
Ro 15-4513, solid