Merck
  • Correlation between the urine profile of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone metabolites and N7-methylguanine in urothelial carcinoma patients.
Correlation between the urine profile of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone metabolites and N7-methylguanine in urothelial carcinoma patients.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2008-12-10)
Hui-Ling Lee, Yu-Mei Hsueh, Chi-Jung Chung, Yeong-Shiau Pu, Louis W Chang, Dennis Paul Hsientang Hsieh, Saou-Hsing Liou, Pinpin Lin
ABSTRACT
A major carcinogen, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK), is present in cigarette smoke and its metabolite, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL), is used as an exposure biomarker for environmental tobacco smoke (ETS). This metabolite (NNAL) can be either detoxified into glucuronidated NNAL (NNAL-Gluc) or activated into an unstable reactive metabolite that methylates DNA along with formation of 4-hydroxy-4-(3-pyridyl)-butyric acid [hydroxy acid (HA)]. Therefore, the carcinogenic risk associated with ETS exposure is greatly modulated by individual variations in metabolic activation and detoxification capabilities. In this study, we defined the urinary HA/total NNAL [HA/total NNAL] ratio as the activation index and NNAL-Gluc/free NNAL [(total NNAL-free NNAL)/free NNAL] ratio as the detoxification index of NNK. The major methylated DNA adduct N(7)-methylguanine (N(7)-MeG), considered as the carcinogenic biomarker for cigarette smoking, was excreted in urine. The objective of this study was to investigate the effects of these metabolic indexes of NNK on N(7)-MeG urinary excretion in a population of urothelial carcinoma patients. Urinary levels of total NNAL (free NNAL plus NNAL-Gluc), free NNAL, HA, and N(7)-MeG were positively correlated with smoking. Furthermore, activation index and detoxification index correlated positively and negatively with N(7)-MeG levels, respectively. Our results suggest that these metabolic indices may represent the phenotype of individual metabolism capability and modulate the carcinogenic risk of ETS exposure.
Sigma-Aldrich
7-Methylguanine, ≥98.0% (HPLC)