Merck
  • A straightforward alpha-selective aromatic glycosylation and its application for stereospecific synthesis of 4-methylumbelliferyl alpha-T-antigen.
A straightforward alpha-selective aromatic glycosylation and its application for stereospecific synthesis of 4-methylumbelliferyl alpha-T-antigen.
Carbohydrate research (2009-01-10)
Shih-Sheng Chang, Chun-Cheng Lin, Yaw-Kuen Li, Kwok-Kong Tony Mong
ABSTRACT
A practical and efficient alpha-selective aromatic glycosylation with simple per-O-acetyl glycopyranosyl trichloroacetimidates is reported. The method is particularly effective for L-fucosyl and 2-azido-2-deoxy-D-galatosaminyl imidates, with which exclusive alpha-selectivity was achieved. The synthetic utility of this method was demonstrated in the stereoselective synthesis of 4-methylumbelliferyl alpha-T-antigen.