Merck
  • Shadows across mu-Star? Constitutively active mu-opioid receptors revisited.
Shadows across mu-Star? Constitutively active mu-opioid receptors revisited.
British journal of pharmacology (2009-04-17)
Mark Connor
ABSTRACT
Constitutively active mu-opioid receptors (mu* receptors) are reported to be formed following prolonged agonist treatment of cells or whole animals. mu* receptors signal in the absence of activating ligand and a blockade of mu* activation of G-proteins by naloxone and naltrexone has been suggested to underlie the profound withdrawal syndrome precipitated by these antagonists in vivo. In this issue of the Journal, Divin et al. examined whether treatment of C6 glioma cells with mu-opioid receptor agonists produced constitutively active mu-opioid receptors or other commonly reported adaptations to prolonged agonist treatment. Adenylyl cyclase superactivation was readily apparent following agonist treatment but there was no evidence of the formation of constitutively active mu-opioid receptors. This result challenges the notion that prolonged agonist exposure inevitably produces mu* receptors, and is consistent with many studies of adaptations in neurons produced by chronic agonist treatment. The investigators provide no explanation of their failure to see mu* receptors in C6 cells, but this is perhaps understandable because the molecular nature of mu* receptors remains elusive, and the precise mechanisms that lead to their formation are unknown. Without knowing exactly what mu* receptors are, how they are formed and how they signal, understanding their role in cellular adaptations to prolonged opioid treatment will remain impossible. Studies such as this should refocus attention on establishing the molecular mechanisms that underlie that phenomenon of mu* receptors.
Product Description
Supelco
6β-Naltrexol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®