Merck
A case of ocular hypertension complicated by SUNCT syndrome.
Annals of ophthalmology (Skokie, Ill.) (2010-12-09)
Junko Murakami, Akio Kim, Takeshi Sugiyama, Keiji Inoue, Chota Matsumoto, Yoshikazu Shimomura
ABSTRACT
We report a 53-year-old woman with laser iridotomy (LI)-resistant angle-closure and conjunctival injection, which was thought to be the cause of ciliochoroidal effusion associated with short-lasting unilateral neuralgiform headache with conjunctival injection and tearing (SUNCT) syndrome. LI had no effect on any of the symptoms except for intraocular pressure. The symptoms disappeared after a subsequent procedure for SUNCT syndrome. MRI of the left eye showed ciliochoroidal effusion at paroxysm and was normalized upon relief.
Sigma-Aldrich
Lomerizine dihydrochloride, ≥98% (HPLC), powder