Merck
  • Chemoselective C-4 aerobic oxidation of catechin derivatives catalyzed by the Trametes villosa laccase/1-hydroxybenzotriazole system: synthetic and mechanistic aspects.
Chemoselective C-4 aerobic oxidation of catechin derivatives catalyzed by the Trametes villosa laccase/1-hydroxybenzotriazole system: synthetic and mechanistic aspects.
The Journal of organic chemistry (2011-01-06)
Roberta Bernini, Fernanda Crisante, Patrizia Gentili, Fabio Morana, Marco Pierini, Monica Piras
ABSTRACT
Catechin derivatives were oxidized in air in the presence of the Trametes villosa laccase/1-hydroxybenzotriazole (HBT) system in buffered water/1,4-dioxane as reaction medium. The oxidation products, flavan-3,4-diols and the corresponding C-4 ketones, are bioactive compounds and useful intermediates for the hemisynthesis of proanthocyanidins, plant polyphenols which provide beneficial health properties for humans. Determinations of oxidation potentials excluded that catechin derivatives could be directly oxidized by laccase Cu(II), while it resulted in the H-abstraction from benzylic positions being promptly promoted by the enzyme in the presence of the mediator HBT, the parent species producing in situ the reactive intermediate benzotriazole-N-oxyl (BTNO) radical. A remarkable and unexpected result for the laccase/HBT oxidative system has been the chemoselective insertion of the oxygen atom into the C-4-H bond of catechin derivatives. Mechanistic aspects of the oxidation reaction have been investigated in detail for the first time in order to corroborate these results. Since the collected experimental findings could not alone provide information useful to clarify the origin of the observed chemoselectivity, these data were expressly supplemented with information derived by suitable molecular modeling investigations. The integrated evaluation of the dissociation energies of the C-H bonds calculated both by semiempirical and DFT methods and the differential activation energies of the process estimated by a molecular modeling approach suggested that the observed selective oxidation at the C-4 carbon has a kinetic origin.