Merck
  • Study of energetics and structure of 1,2,3-benzotriazin-4(3H)-one and its 1H and enol tautomers.
Study of energetics and structure of 1,2,3-benzotriazin-4(3H)-one and its 1H and enol tautomers.
The journal of physical chemistry. B (2011-05-06)
Margarida S Miranda, M Agostinha R Matos, Victor M F Morais, Joel F Liebman
ABSTRACT
This paper reports an experimental and computational study on the energetics of 1,2,3-benzotriazin-4(3H)-one. The standard (p° = 0.1 MPa) molar enthalpy of formation of solid 1,2,3-benzotriazin-4(3H)-one, at T = 298.15 K, was derived from its standard massic energy of combustion measured by static bomb combustion calorimetry in oxygen. The Calvet high-temperature vacuum sublimation technique was used to measure the respective standard molar enthalpy of sublimation at T = 298.15 K. From these two experimentally determined thermodynamic parameters, we have calculated the standard molar enthalpy of formation of 1,2,3-benzotriazin-4(3H)-one in the gas phase at T = 298.15 K, (200.9 ± 3.8) kJ·mol(-1). Interrelations between structure and energy for 1,2,3-benzotriazin-4(3H)-one, the tautomer 1,2,3-benzotriazin-4(1H)-one, and the enol tautomer 1,2,3-benzotriazin-4-ol were discussed based on density functional theory (DFT) calculations with the B3LYP hybrid functional and the 6-311++G(d,p) basis set. The gas-phase enthalpy of formation of 1,2,3-benzotriazin-4(3H)-one was estimated from quantum chemical calculations using the G3(MP2)//B3LYP composite method. Nucleus-independent chemical shifts (NICS) were also calculated with the purpose of analyzing the aromaticity of the benzenic and heterocyclic rings of the title molecule and others related tautomerically to it.