Merck
  • Protein kinase C phosphorylates the cAMP response element binding protein in the hypothalamic paraventricular nucleus during morphine withdrawal.
Protein kinase C phosphorylates the cAMP response element binding protein in the hypothalamic paraventricular nucleus during morphine withdrawal.
British journal of pharmacology (2011-05-28)
F Martín, L Mora, Ml Laorden, Mv Milanés
ABSTRACT
Exposure to drugs of abuse or stress results in adaptation in the brain involving changes in gene expression and transcription factors. Morphine withdrawal modulates gene expression through various second-messenger signal transduction systems. Here, we investigated changes in activation of the transcription factor, cAMP-response element binding protein (CREB), in the hypothalamic paraventricular nucleus (PVN) and the kinases that may mediate the morphine withdrawal-triggered activation of CREB and the response of the hypothalamic-pituitary-adrenocortical (HPA) axis after naloxone-induced morphine withdrawal. The effects of morphine dependence and withdrawal, phosphorylated CREB (pCREB), corticotrophin-releasing factor (CRF) expression in the PVN and HPA axis activity were measured using immunoblotting, immunohistochemistry and radioimmunoassay in controls and in morphine-dependent rats, withdrawn with naloxone and pretreated with vehicle, calphostin C, chelerythrine (inhibitors of protein kinase C (PKC) or SL-327 [inhibitor of extracellular signal regulated kinase (ERK) kinase]. In addition, changes in PKCα and PKCγ immunoreactivity were measured after 60 min of withdrawal. In morphine-withdrawn rats, pCREB immunoreactivity was increased within CRF immunoreactive neurons in the PVN and plasma corticosterone levels were raised. SL-327, at doses that reduced the augmented pERK levels in the PVN, did not attenuate the rise in pCREB immunoreactivity or plasma corticosterone secretion. In contrast, PKC inhibition reduced the withdrawal-triggered rise in pCREB, pERK1/2 and corticosterone secretion. PKC mediated, in part, both CREB activation and the HPA response to morphine withdrawal. The ERK kinase/ERK pathway might not be necessary for either activation of CREB or HPA axis hyperactivity.
Sigma-Aldrich
Chelerythrine chloride, ≥95% (TLC), powder
Sigma-Aldrich
Calphostin C from Cladosporium cladosporioides, ≥90% (HPLC), powder
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder