Merck
  • Drug screening of whole blood by ultra-performance liquid chromatography-tandem mass spectrometry.
Drug screening of whole blood by ultra-performance liquid chromatography-tandem mass spectrometry.
Journal of analytical toxicology (2011-05-31)
Elisabeth Leere Oiestad, Unni Johansen, Ase Marit Leere Oiestad, Asbjørg Solberg Christophersen
ABSTRACT
An ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS-MS) method for screening of drugs in whole blood has been developed and validated. Samples were prepared by supported liquid-liquid extraction on ChemElute(®) columns with ethyl acetate/heptane (4:1). LC separation was achieved with an Acquity HSS T3-column (2.1 100 mm, 1.8-μm particle). Mass detection was performed by positive ion mode electrospray MS-MS and included the following drugs/metabolites: morphine, codeine, ethyl morphine, oxycodone, buprenorphine, methadone, cocaine, methylphenidate, amphetamine, methamphetamine, 3,4-methylenedioxymethamphetamine (MDMA), Δ(9)-tetrahydrocannabinol (THC), fentanyl, alprazolam, bromazepam, clonazepam, diazepam, nordiazepam, 3-OH-diazepam, fenazepam, flunitrazepam, lorazepam, nitrazepam, oxazepam, zopiclone, zolpidem, carisoprodol, and meprobamate. The cycle time was 9 min, and within- and between-day relative coefficients of variation varied from 1% to 33% and 2% to 58%, respectively. Extraction recoveries from whole blood were > 50% except for morphine and THC. The limit of quantitation was 0.1 to 521 ng/mL, depending on the drug.
Product Description
Supelco
Ethylmorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®