Merck
  • Air- and heat-stable planar tri-p-quinodimethane with distinct biradical characteristics.
Air- and heat-stable planar tri-p-quinodimethane with distinct biradical characteristics.
Journal of the American Chemical Society (2011-09-22)
Xiaozhang Zhu, Hayato Tsuji, Koji Nakabayashi, Shin-ichi Ohkoshi, Eiichi Nakamura
ABSTRACT
A heptacyclic carbocycle possessing three p-quinodimethane units conjugated in one plane has been synthesized and shown to exhibit distinct biradical characteristics. The molecule has a HOMO/LUMO band gap of ca. 1 eV and a S(0)-T(1) energy gap of 2.12 kcal/mol, and it absorbs and emits near-IR light at room temperature. It is air-stable under ambient light for several months and thermally stable up to 160 °C under nitrogen, and it undergoes reversible two-electron oxidation and reduction. The synthetic approach is such that a smaller and larger oligo-p-quinodimethane can be synthesized.