Merck
  • Laccase-mediated system pretreatment to enhance the effect of hydrogen peroxide bleaching of cotton fabric.
Laccase-mediated system pretreatment to enhance the effect of hydrogen peroxide bleaching of cotton fabric.
International journal of biological macromolecules (2011-12-14)
Liqiang Tian, C Branford-White, Wen Wang, Huali Nie, Limin Zhu
ABSTRACT
This study evaluates the bleaching efficiency of the hydrogen peroxide bleaching process combined with laccase-mediated system pretreatment (LMS-HPBP) in the treatment of scoured cotton fabric. By changing the factors of laccase-mediated system pretreatment and the hydrogen peroxide bleaching process and examining the subsequent whiteness value and retained tensile strength of the samples, we find three LMS-HPBP processes that are more environment friendly than the conventional hydrogen peroxide bleaching process (CHPBP): (i) bleaching with lower dosage of hydrogen peroxide; (ii) bleaching at reduced temperature; (iii) bleaching for shortened duration. Whiteness, retained tensile strength and K/S values of cotton fabric samples treated by i-iii processes were similar to or higher than those by CHPBP. X-ray diffraction (XRD) analysis also demonstrated that the three processes rendered fabric of both lower crystallinity and bigger crystallite size than those by CHPBP. In addition, the "green" short-flow process was developed to treat cotton fabric and the results obtained shows this method is feasible as a new energy-saving process.