Merck
A novel series of pyrazolylpiperidine N-type calcium channel blockers.
Bioorganic & medicinal chemistry letters (2012-05-23)
Nalin L Subasinghe, Mark J Wall, Michael P Winters, Ning Qin, Mary Lou Lubin, Michael F A Finley, Michael R Brandt, Michael P Neeper, Craig R Schneider, Raymond W Colburn, Christopher M Flores, Zhihua Sui
ABSTRACT
Selective blockers of the N-type calcium channel have proven to be effective in animal models of chronic pain. However, even though intrathecally delivered synthetic ω-conotoxin MVIIA from Conus magnus (ziconotide [Prialt®]) has been approved for the treatment of chronic pain in humans, its mode of delivery and narrow therapeutic window have limited its usefulness. Therefore, the identification of orally active, small-molecule N-type calcium channel blockers would represent a significant advancement in the treatment of chronic pain. A novel series of pyrazole-based N-type calcium channel blockers was identified by structural modification of a high-throughput screening hit and further optimized to improve potency and metabolic stability. In vivo efficacy in rat models of inflammatory and neuropathic pain was demonstrated by a representative compound from this series.