Merck
  • The hepatoprotection of caffeic acid and rosmarinic acid, major compounds of Perilla frutescens, against t-BHP-induced oxidative liver damage.
The hepatoprotection of caffeic acid and rosmarinic acid, major compounds of Perilla frutescens, against t-BHP-induced oxidative liver damage.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2013-01-12)
Sung-Yong Yang, Chung-Oui Hong, Gung Pyo Lee, Cheong-Tae Kim, Kwang-Won Lee
ABSTRACT
Perilla frutescens leaves are often used in East Asian gourmet food. In this study, we investigated the hepatoprotective effects of caffeic acid (CA), rosmarinic acid (RA), and their combination. P. frutescens contains 1.32μg CA/mg dry material (DM) and 26.84μg RA/mg DM analyzed by HPLC-DAD and HPLC-MS. CA remarkably reduced the oxidative damage than rosmarinic acid in an in vitro study. Oral intubation with CA or RA alone for five days was conducted prior to treatment with a single dose of tert-butyl hydroperoxide (0.5mmol/kg b.w., i.p.), which led to a significant reduction of indicators of hepatic toxicity, such as aspartate aminotransferase, alanine aminotransferase, oxidized glutathione, lipid peroxidation and enzyme activities related to antioxidant such as catalase, glutathione peroxidase and superoxide dismutase. Interestingly, compared to treatment with CA or RA alone, a combination of both compounds more increased the endogenous antioxidant enzymes and glutathione (GSH) and decreased lipid peroxidation in livers. These results suggest that CA from perilla leaves plays a role in the increased hepatic GSH concentration, and shows an additive hepatic protection with RA against oxidative hepatic damage.
Product Description
Supelco
trans-Caffeic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
Rosmarinic acid, 96%
Rosmarinic acid, primary reference standard
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)