Merck
  • Bacterial contamination of stethoscope chest pieces and the effect of daily cleaning.
Bacterial contamination of stethoscope chest pieces and the effect of daily cleaning.
Journal of veterinary internal medicine (2013-02-22)
H Fujita, B Hansen, R Hanel
ABSTRACT
Stethoscopes are a potential source of nosocomial infection for hospitalized humans, a phenomenon not previously studied in companion animals. To determine if daily cleaning of stethoscope chest pieces reduces bacterial contamination between cleanings. Client-owned dogs and cats. Prospective observational study. In phase 1, bacterial cultures were obtained from the chest pieces of 10 participant stethoscopes once weekly for 3 weeks. In phase 2, stethoscopes were cleaned daily and 2 culture samples were obtained once weekly, immediately before and after cleaning with 70% isopropyl alcohol, for 3 weeks. Daily cleaning eliminated bacteria immediately after each cleaning (P = .004), but did not reduce the rate of positive cultures obtained before cleaning in phase 2. Cultures were positive for 20/30 (67%) samples during phase 1 and 18/30 (60%) obtained before daily cleaning during phase 2. Recovered organisms included normal skin flora, agents of opportunistic infections, and potential pathogens. The only genus that was repeatedly recovered from the same stethoscope for 2 or more consecutive weeks was Bacillus sp. Daily cleaning was highly effective at removing bacteria, but provided no reduction in precleaning contamination. Cleaning stethoscopes after use on dogs or cats infected with pathogenic bacteria and before use on immunocompromised animals should be considered.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Isopropanol, 70% in H2O
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications