Merck
  • Mixed sugar-core-phosphate chelation of D-fructose 1,6-bisphosphate with the Re(V)O(tmen) metal fragment.
Mixed sugar-core-phosphate chelation of D-fructose 1,6-bisphosphate with the Re(V)O(tmen) metal fragment.
Dalton transactions (Cambridge, England : 2003) (2013-03-02)
Martin Steinborn, Mihael Suhanji, Peter Klüfers
ABSTRACT
Sugar phosphates provide metal-binding sites both at their sugar core and at their phosphate group(s). Mixed sugar-core-phosphate chelation has been considered as a typical bonding mode within the physiological pH range for the central metabolite D-fructose 1,6-bisphosphate. The Re(V)O(tmen) metal fragment was used to enrich this coordination type. The formation of the [ReO(tmen)(Fruf2,3H-21,6P2H2-κ(3)O(2,3,P1))](-) monoanion was determined by NMR spectroscopy and mass spectrometry. The model compound rac-glycerol 1-phosphate yielded similar results in terms of NMR spectroscopy. Crystal-structure analyses of [ReO(tmen)(rac-Glyc2,3H-21PH-κ(3)O(2,3,P))]·2H2O and [ReO(phen)(rac-Glyc2,3H-21PH-κ(3)O(2,3,P))]·MeOH confirmed the coordination pattern.
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis