Merck
  • Selective syntheses of Δ(α,β) and Δ(β,γ) butenolides from allylic cyclopropenecarboxylates via tandem ring expansion/[3,3]-sigmatropic rearrangements.
Selective syntheses of Δ(α,β) and Δ(β,γ) butenolides from allylic cyclopropenecarboxylates via tandem ring expansion/[3,3]-sigmatropic rearrangements.
Organic letters (2013-03-22)
Xiaocong Xie, Yi Li, Joseph M Fox
ABSTRACT
Allylic cyclopropenecarboxylates undergo ring expansion reactions to give 2-allyloxyfuran intermediates, which subsequently rearrange to Δ(β,γ) butenolides via a Claisen rearrangement or to the corresponding Δ(α,β) butenolides via further Cope rearrangement. Also described are methods for chirality transfer in the rearrangement of nonracemic allylic esters.
Sigma-Aldrich
2(5H)-Furanone, 98%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis