Merck
  • Biomimetic synthesis of zeylanone and zeylanone epoxide by dimerization of 2-methyl-1,4-naphthoquinone.
Biomimetic synthesis of zeylanone and zeylanone epoxide by dimerization of 2-methyl-1,4-naphthoquinone.
Organic letters (2013-03-27)
Sayako Maruo, Kazuyuki Nishio, Takahiro Sasamori, Norihiro Tokitoh, Kouji Kuramochi, Kazunori Tsubaki
ABSTRACT
A biomimetic synthesis of zeylanone and zeylanone epoxide, which are natural dimeric naphthoquinones, has been accomplished starting from plumbagin, a natural monomeric naphthoquinone. The key features of our synthesis are cascade intermolecular and intramolecular Michael reactions, followed by epoxidation of the resultant hydroquinone with molecular oxygen.
Product Description
Supelco
Menadione (K3), analytical standard
Menadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications