- Catalytic deoxydehydration of glycols with alcohol reductants.
Catalytic deoxydehydration of glycols with alcohol reductants.
ChemSusChem (2013-03-28)
Camille Boucher-Jacobs, Kenneth M Nicholas
PMID23532937
ABSTRACT
Top shelf dehydration: Ammonium perrhenate catalysts combined with benzylic alcohol reductants are used for the efficient deoxydehydration of glycols to olefins. The olefin and aldehyde products can be easily separated and isolated. It is also demonstrated that the catalyst can be recovered and reused because of its low solubility in aromatic solvents.
Product Description
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard