Merck
  • Nearly degenerate wavelength-multiplexed polarization entanglement by cascaded optical nonlinearities in a PPLN ridge waveguide device.
Nearly degenerate wavelength-multiplexed polarization entanglement by cascaded optical nonlinearities in a PPLN ridge waveguide device.
Optics express (2013-04-03)
Shin Arahira, Hitoshi Murai
ABSTRACT
In this paper we report the generation of wavelength-multiplexed polarization-entangled photon pairs in the 1.5-μm communication wavelength band by using cascaded optical second nonlinearities (sum-frequency generation and subsequent spontaneous parametric down-conversion, c-SFG/SPDC) in a periodically poled LiNbO(3) ridge waveguide device. The c-SFG/SPDC method makes it possible to fully use the broad spectral bandwidth of SPDC in nearly frequency-degenerate conditions, and can provide more than 50 pairs of wavelength channels for the entangled photon pairs in the 1.5-μm wavelength band, using only standard optical resources in the telecom field. Visibilities higher than 98% were clearly observed in two-photon interference fringes for all the wavelength channels under investigation (eight pairs). We further performed a detailed experimental investigation of the cross-talk characteristics and the impact of detuning the pump wavelengths.
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil