Merck
  • Valproic acid-induced hyperammonaemic coma and unrecognised portosystemic shunt.
Valproic acid-induced hyperammonaemic coma and unrecognised portosystemic shunt.
Epileptic disorders : international epilepsy journal with videotape (2013-06-19)
Hipólito Nzwalo, Leonor Carrapatoso, Fátima Ferreira, Carlos Basilio
ABSTRACT
Hyperammonaemic encephalopathy is a rare and potentially fatal complication of valproic acid treatment. The clinical presentation of hyperammonaemic encephalopathy is wide and includes seizures and coma. We present a case of hyperammonaemic coma precipitated by sodium valproate use for symptomatic epilepsy in a patient with unrecognised portosystemic shunt, secondary to earlier alcoholism. The absence of any stigmata of chronic liver disease and laboratory markers of liver dysfunction delayed the recognition of this alcohol-related complication. The portal vein bypass led to a refractory, valproic acid-induced hyperammonaemic coma. The patient fully recovered after dialysis treatment.
Product Description
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid