Merck
  • The lack of L-PG production and the repercussions of it in regards to M. Tuberculosis interactions with mononuclear phagocytes.
The lack of L-PG production and the repercussions of it in regards to M. Tuberculosis interactions with mononuclear phagocytes.
Acta microbiologica et immunologica Hungarica (2013-07-06)
Marek Fol, Anna Głobińska, Paweł Stączek, Magdalena Kowalewicz-Kulbat, Magdalena Druszczyńska, Murty V V S Madiraju, Wiesława Rudnicka
ABSTRACT
The lysine connection with phosphatidylglycerol (PG) alters the M. tuberculosis(Mtb) surface charge, and consequently it may decrease the bacterial vulnerability to antimicrobial action of the immune cells. The aim of the study was to assess the significance of PG lysinylation in the Mtb interactions with mononuclear phagocytes. Both the Mtb strain with deletion of lysX gene (Mtb-lysX) which is responsible for PG lysinylation as well as the complemented strain (Mtb-compl) was used to infect human blood monocytes or THP-1 cells. The monocytes were obtained by MACS technique, or THP-1 cells. The Mtb-lysX strain has exhibited the enhanced sensitivity to HNP 1-3. However, it was not susceptible to bactericidal action of cathepsin G. The LysX deletion did not influence the Mtb ability of monocyte induction to IL-10 secretion. The intra- and extracellular expression of MHC-II was similarly reduced after the Mtb-lysX or Mtb-Rv infections. Noticeably significant is that the Mtb strain with deleted lysX has not affected the intensity of the gene expression of cathepsin G compared to the uninfected monocytes. That is the clear contrast to what the Mtb-Rv strain has proved. The obtained results suggest that the Mtb ability to lysinylate PG is a participatory element in mycobacterial strategy of survival inside phagocytic cells. However, the extended studies are needed to determine its influence on the other immune cells and define its role in the developing of Mtb infection.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland