Merck
  • Implant abutment screw reverse torque values before and after plasma cleaning.
Implant abutment screw reverse torque values before and after plasma cleaning.
The International journal of prosthodontics (2013-07-10)
Costanza Micarelli, Luigi Canullo, Paolo Baldissara, Marco Clementini
ABSTRACT
This in vitro study analyzed the reverse torque (RevT) of abutment screws following different cleaning treatments. A convenience sample of 50 customized titanium abutment screw complexes was divided into five groups: cleaning by steam (control group), cleaning by Argon plasma (test groups 1 and 2 [with chlorhexidine gel]), and replacement of old screws with new ones (test groups 3 and 4 [with chlorhexidine gel]). Abutments were screwed onto implants and tested for RevT. The RevT of the test groups was significantly higher than that of the control group. No statistically significant difference between test groups was noted except between groups 2 and 3.
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorhexidine, ≥99.5%