Merck
  • 6-Methylpurine, 6-methyl-9-beta-D-ribofuranosylpurine, and 6-hydroxymethyl-9-beta-D-ribofuranosylpurine as antiviral metabolites of Collybia maculata (Basidiomycetes).
6-Methylpurine, 6-methyl-9-beta-D-ribofuranosylpurine, and 6-hydroxymethyl-9-beta-D-ribofuranosylpurine as antiviral metabolites of Collybia maculata (Basidiomycetes).
Zeitschrift fur Naturforschung. C, Journal of biosciences (1987-04-01)
K Leonhardt, T Anke, E Hillen-Maske, W Steglich
ABSTRACT
6-Methylpurine, 6-methyl-9-beta-D-ribofuranosylpurine and 6-hydroxymethyl-9-beta-D-ribofuranosylpurine were isolated from mycelial cultures of Collybia maculata and their antifungal, cytotoxic and antiviral activities investigated. This is the first report on the natural occurrence of these compounds.
Sigma-Aldrich
6-Methylpurine, ≥99%